Skip to Content
Merck
All Photos(1)

Documents

757233

Sigma-Aldrich

2,4-Bis[4-(N,N-diphenylamino)-2,6-dihydroxyphenyl]squaraine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H28N2O6
CAS Number:
Molecular Weight:
632.66
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

mp

328-333 °C

λmax

720 nm

Orbital energy

HOMO 5.3 eV 
LUMO 3.8 eV 

OPV Device Performance

ITO/MoO3/DP-SQ/C60/PTCBI/Ag

  • Short-circuit current density (Jsc): 7.2 mA/cm2
  • Open-circuit voltage (Voc): 0.91 V
  • Fill Factor (FF): 0.7
  • Power Conversion Efficiency (PCE): 4.8 %

SMILES string

Oc1cc(cc(O)c1[C]2[C+]([O-])[C]([C+]2[O-])c3c(O)cc(cc3O)N(c4ccccc4)c5ccccc5)N(c6ccccc6)c7ccccc7

InChI

1S/C40H28N2O6/c43-31-21-29(41(25-13-5-1-6-14-25)26-15-7-2-8-16-26)22-32(44)35(31)37-39(47)38(40(37)48)36-33(45)23-30(24-34(36)46)42(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-24,43-46H

InChI key

FXAALAVLRJSKEQ-UHFFFAOYSA-N

General description

2,4-Bis[4-(N,N-diphenylamino)-2,6-dihydroxyphenyl]squaraine (DPSQ) is a symmetric squaraine that can be used as a green light absorbing donor. The characteristic absorption peak is at 720 nm. It can be used for a variety of electrochemical applications.

Application

DPSQ based dyes can be used for potential application in polymeric solar cells (PSCs), and organic photovoltaic cells (OPVs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and disorder in squaraine-C60 organic solar cells: a theoretical description of molecular packing and electronic coupling at the donor-acceptor interface
Fu Y, et al.
Advances in Functional Materials, 24(24), 3790-3798 (2014)
Control of interface order by inverse quasi-epitaxial growth of squaraine/fullerene thin film photovoltaics
Zimmerman JD, et al.
ACS Nano, 7(10), 9268-9275 (2013)
Functionalized squaraine donors for nanocrystalline organic photovoltaics
Wei G, et al.
ACS Nano, 6(1), 972-978 (2011)
Independent control of bulk and interfacial morphologies of small molecular weight organic heterojunction solar cells
Zimmerman JD, et al.
Nano Letters, 12(8), 4366-4371 (2012)
Quaternary organic solar cells enhanced by cocrystalline squaraines with power conversion efficiencies> 10%
Goh T, et al.
Advanced Energy Materials, 6(21), 1600660-1600660 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service