Skip to Content
Merck
All Photos(1)

Key Documents

753939

Sigma-Aldrich

DBP

98% (HPLC)

Synonym(s):

5,10,15,20-Tetraphenylbisbenz[5,6]indeno[1,2,3-cd:1′,2′,3′-lm]perylene, Dibenzo{[f,f′]-4,4′,7,7′-tetraphenyl}diindeno[1,2,3-cd:1′,2′,3′-lm]perylene, Red 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H36
CAS Number:
Molecular Weight:
804.97
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

98% (HPLC)

form

solid

loss

0.5 wt. %, 373 °C

mp

>400 °C

Orbital energy

HOMO 5.5 eV (CV)
LUMO 3.5 eV (CV)

OPV Device Performance

ITO/DBP/C60/BCP/Al

  • Short-circuit current density (Jsc): 6.3 mA/cm2
  • Open-circuit voltage (Voc): 0.92 V
  • Fill Factor (FF): 0.62
  • Power Conversion Efficiency (PCE): 0.36 %

SMILES string

c1ccc(cc1)-c2c-3c(-c4ccc5c6ccc7-c8c(-c9ccc(c%10ccc-3c4c5%10)c6c79)c(-c%11ccccc%11)c%12ccccc%12c8-c%13ccccc%13)c(-c%14ccccc%14)c%15ccccc2%15

InChI

1S/C64H36/c1-5-17-37(18-6-1)53-41-25-13-14-26-42(41)54(38-19-7-2-8-20-38)62-50-34-30-46-48-32-36-52-60-51(35-31-47(58(48)60)45-29-33-49(61(53)62)59(50)57(45)46)63-55(39-21-9-3-10-22-39)43-27-15-16-28-44(43)56(64(52)63)40-23-11-4-12-24-40/h1-36H

InChI key

WPPDXAHGCGPUPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kamin J Johnson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 105(2), 360-367 (2008-07-16)
Phenotype results from interactions between genetics and environment, but for most environmental chemical exposures, such interactions are theoretical. The phenotypic response of the testis to in utero dibutyl phthalate (DBP) exposure was compared between two strains of Long-Evans (LE) rats
Simona Perga et al.
PloS one, 10(6), e0129291-e0129291 (2015-06-06)
Multiple sclerosis (MS) is a multifactorial autoimmune disease of the central nervous system with a heterogeneous and unpredictable course. To date there are no prognostic biomarkers even if they would be extremely useful for early patient intervention with personalized therapies.
Lukas D Trachsel et al.
Journal of hypertension, 33(6), 1276-1283 (2015-02-11)
Extensive endurance training and arterial hypertension are established risk factors for atrial fibrillation. We aimed to assess the proportion of masked hypertension in endurance athletes and the impact on cardiac remodeling, mechanics, and supraventricular tachycardias (SVT). Male participants of a
Jonatan Axelsson et al.
Environment international, 85, 54-60 (2015-09-01)
In animals, exposure to certain phthalates negatively affects the male reproductive function. Human results are conflicting and mostly based on subfertile males, in whom the association between exposure and reproductive function may differ from the general population. To study if
Sheng-Xian Wu et al.
Current vascular pharmacology, 13(4), 504-519 (2014-11-02)
There are more than 300 million patients with hypertension in China, and at least 1 in 5 is using or has ever used Chinese Medicine (CM) treatment. This article reviews the efficacy and safety of CM as monotherapy and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service