Skip to Content
Merck
All Photos(1)

Documents

753637

Sigma-Aldrich

Gold nanoparticles

40 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

nanoparticles
suspension

concentration

~7.2E+10 particles/mL

OD

1

diameter

40 nm

λmax

526-531 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles (AuNPs), 40 nm are quasi dynamic colloidal nanomaterials with good surface chemistry and high surface to volume ratio. Their properties include high stability, biocompatibility, and low toxicity which make them useful in biological applications.

Application

AuNPs can be used for a variety of applications such as biomedical, electrochemical and energy based devices. They can also be used in drug delivery and biosensing based device fabrication.

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold Nanoparticles For Physics, Chemistry And Biology (Second Edition) (2012)
Guy Nir et al.
PLoS genetics, 14(12), e1007872-e1007872 (2018-12-27)
Chromosome organization is crucial for genome function. Here, we present a method for visualizing chromosomal DNA at super-resolution and then integrating Hi-C data to produce three-dimensional models of chromosome organization. Using the super-resolution microscopy methods of OligoSTORM and OligoDNA-PAINT, we
Gold nanoparticles for applications in cancer radiotherapy: Mechanisms and recent advancements
Her S, et al.
Advanced Drug Delivery Reviews, 109(6), 84-101 (2017)
Gold nanoparticles for applications in energy and environment: synthesis and characterization
Chen Y
Rare Metals, 30(1), 116-120 (2011)
Spectroscopic Study of the Interaction of Carboxyl-Modified Gold Nanoparticles with Liposomes of Different Chain Lengths and Controlled Drug Release by Layer-by-Layer Technology
Kanwa N, et al.
The Journal of Physical Chemistry B, 121(50), 11333-11343 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service