Skip to Content
Merck
All Photos(1)

Key Documents

74860

Sigma-Aldrich

(±)-2-Octanol

≥96.0% (GC)

Synonym(s):

Hexyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719325
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥96.0% (GC)
bp:
179-180 °C

Quality Level

Assay

≥96.0% (GC)

form

liquid

refractive index

n20/D 1.426

bp

179-180 °C

density

0.819 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Yamamoto et al.
Applied microbiology and biotechnology, 97(18), 8087-8096 (2013-01-01)
A novel enantioselective alcohol dehydrogenase, (R)-2-octanol dehydrogenase (PfODH), was discovered among methylotrophic microorganisms. The enzyme was purified from Pichia finlandica and characterized. The molecular mass of the enzyme was estimated to be 83,000 and 30,000 by gel filtration and sodium
G D Rees et al.
Biochimica et biophysica acta, 1073(3), 493-501 (1991-04-09)
Lipase from three different sources has been immobilised in microemulsion-based gels (MBGs) with retention of catalytic activity. Such lipase-containing MBGs prove to be novel solid-phase catalysts for use in apolar organic solvents such as n-heptane. Using these systems, preparative-scale synthesis
J S Schweitzer et al.
Journal of neurophysiology, 84(2), 927-933 (2000-08-12)
Under conditions of low [Ca(2+)](o) and high [K(+)](o), the rat dentate granule cell layer in vitro develops recurrent spontaneous prolonged field bursts that resemble an in vivo phenomenon called maximal dentate activation. To understand how pH changes in vivo might
Chi-Yu Huang et al.
International journal of pharmaceutics, 325(1-2), 132-139 (2006-07-22)
Core-shell type of nanoparticles (NPs) with manipulated degradation rate and balanced hydrophilic/hydrophobic properties were designed and characterized. The NPs based on the copolymers of n-butyl cyanoacrylate (BCA) and 2-octyl cyanoacrylate (OCA) were prepared by anion emulsion polymerization in 0.01N HCl
Deracemization of secondary alcohols through a concurrent tandem biocatalytic oxidation and reduction.
Constance V Voss et al.
Angewandte Chemie (International ed. in English), 47(4), 741-745 (2007-12-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service