747467
Iron oxide(II,III) magnetic nanopowder
30 nm avg. part. size (TEM), NHS ester functionalized
Synonym(s):
Fe NP NHS, FexOy
About This Item
Recommended Products
form
nanopowder
powder
composition
Fe, ~1.25 % (w/w)
magnetization
>45 emu/g, at room temperature; under 4500 Oe
color
light brown to dark brown, and Dark Grey
avg. part. size
30 nm (TEM)
functional group
NHS ester (functionalized)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service