Skip to Content
Merck
All Photos(1)

Key Documents

742058

Sigma-Aldrich

Gold nanoparticles

150 nm diameter, stabilized suspension in citrate buffer

Synonym(s):

Au NP, Gold Colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

contains

Proprietary Surfactant as stabilizer

concentration

~3.6E+9 particles/mL

diameter

150 nm

PDI

<0.2

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications. Note: This product is not ideal for protein adsorption. For gold nanoparticles suitable for adsorption of proteins and other ligands for conjugate development, please see our gold nanoparticles stabilized in PBS.

This material is highly monodisperse (<12% variability in size and shape), and provides significantly improved surface reactivity. Applications include Surface Enhanced Raman Lables, Sensing/Detection, Biological Targeting, Plasmonics and Electronics.

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service