Skip to Content
Merck
All Photos(1)

Key Documents

737038

Sigma-Aldrich

3-Aminotetrahydrofuran

95%

Synonym(s):

Tetrahydrofuran-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.458

density

1.012 g/mL at 25 °C

SMILES string

NC1CCOC1

InChI

1S/C4H9NO/c5-4-1-2-6-3-4/h4H,1-3,5H2

InChI key

MIPHRQMEIYLZFZ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F

Flash Point(C)

48.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service