Skip to Content
Merck
All Photos(1)

Key Documents

730300

Sigma-Aldrich

Methacrylic acid N-hydroxysuccinimide ester

98%

Synonym(s):

N-(Methacryloxy)succinimide, N-(Methacryloyloxy)succinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO4
CAS Number:
Molecular Weight:
183.16
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

101-105 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)ON1C(=O)CCC1=O

InChI

1S/C8H9NO4/c1-5(2)8(12)13-9-6(10)3-4-7(9)11/h1,3-4H2,2H3

InChI key

ACGJEMXWUYWELU-UHFFFAOYSA-N

General description

Methacrylic acid N-hydroxysuccinimide ester(NHS-MA) belongs to the class of reactive monomers known as N-hydroxysuccinimide (NHS) esters. It is commonly used for functionalizing biomolecules through amine-reactive coupling reactions infields such as bioconjugation, protein labeling, and peptide synthesis. It also serves as a crosslinking agent or linker molecule for the conjugation of drugs or targeting ligands to carrier materials.

Application

Methacrylic acid N-hydroxysuccinimide ester can be used:
  • As a monomer to prepare degradable amphiphilic diblock copolymer microparticles via RAFT polymerization, for low pH-triggered drug delivery. NHS-MA can shield the drug molecule from degradation, enhance its solubility, and improve its pharmacokinetic properties.
  • For the surface functionalization of poly-ε-caprolactone (PCL) scaffolds used for tissue engineering. NHS groups are used to couple with chitosan of various molecular weights.
  • To prepare biocompatible polymer hydrogel for enzymatic biofuel cells. The hydrogel can serve as an enzyme-immobilizing matrix for enzymatic bioelectrodes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrosprayed cysteine-functionalized degradable amphiphilic block copolymer microparticles for low pH-triggered drug delivery
Marie Finnegan, et al
Introduction to Polymer Chemistry, 10 (2019)
Chitosan functionalized poly-?-caprolactone electrospun fibers and 3D printed scaffolds as antibacterial materials for tissue engineering applications
Myriam G. Tardajos, et al.
Carbohydrate Polymers, 191, 127-135 (2018)
Postpolymerization modification of poly (pentafluorophenyl methacrylate): Synthesis of a diverse water-soluble polymer library
Gibson MI, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(17), 4332-4345 (2009)
A Modifiable, Spontaneously Formed Polymer Gel with Zwitterionic and N-Hydroxysuccinimide Moieties for an Enzymatic Biofuel Cell
Yixuan Huang, et al.
ACS applied polymer materials, 3, 631-639 (2021)
Controlled polymerization of N-isopropylacrylamide with an activated methacrylic ester
Savariar EN and Thayumanavan S
Journal of Polymer Science Part A: Polymer Chemistry, 42(24), 6340-6345 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service