Skip to Content
Merck
All Photos(1)

Key Documents

253375

Sigma-Aldrich

3-Maleimido-PROXYL

free radical

Synonym(s):

3-Maleimido-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N2O3
CAS Number:
Molecular Weight:
237.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

mp

111-113 °C (lit.)

functional group

imide
maleimide

storage temp.

2-8°C

SMILES string

CC1(C)CC(N2C(=O)C=CC2=O)C(C)(C)N1[O]

InChI

1S/C12H17N2O3/c1-11(2)7-8(12(3,4)14(11)17)13-9(15)5-6-10(13)16/h5-6,8H,7H2,1-4H3

InChI key

HGNHBHXFYUYUIA-UHFFFAOYSA-N

Application

Spin label used for study of local protein structure and for determining molecular changes in erythrocyte membranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Tseng et al.
Journal of food science, 77(9), H192-H201 (2012-08-23)
The effects of different drying methods (40 °C conventional and vacuum oven, 25 °C ambient air and freeze dry) on the stability of two red wine grape (Pinot Noir, PN and Merlot, M) byproducts, pomace containing skins and seeds (P)
H Dugas et al.
Canadian journal of biochemistry, 57(12), 1407-1415 (1979-12-01)
We have used a series of N-(1-oxyl-2,2,5,5-tetramethyl-3-pyrrolidinyl) maleimide spin labels of different length to label, covalently and selectively, the most reactive sulfhydryl groups of 70S ribosomal proteins of Escherichia coli. Under short periods of labeling (1--2 min), less than two
Oleg Y Dmitriev et al.
Biochimica et biophysica acta, 1777(2), 227-237 (2008-01-08)
Subunit a in the membrane traversing F0 sector of Escherichia coli ATP synthase is known to fold with five transmembrane helices (TMHs) with residue 218 in TMH IV packing close to residue 248 in TMH V. In this study, we
L Zhao et al.
Biophysical journal, 69(3), 994-999 (1995-09-01)
Electron paramagnetic resonance spectroscopy of a spin probe attached to cys-707 on myosin cross-bridges was used to monitor the orientation of the myosin catalytic domain at the beginning and end of the working power stroke in active muscle. Elevated concentrations
P D Morse
Biochimica et biophysica acta, 844(3), 337-345 (1985-03-21)
In a recent paper, Daveloose et al. (Daveloose, D., Fabre, G., Berleur, F., Testylier, G. and Letterrier, F. (1983) Biochim. Biophys. Acta 763, 41-49) described a technique to measure the internal microviscosity of erythrocytes using the spin label MAL-3 (2,2,5,5-tetramethyl-3-malimidopyrrolidinyl-N-oxyl)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service