728551
Bis[(2-dimethylamino)phenyl]amine nickel(II) chloride
≥97% (AT)
Synonym(s):
Chloro[N2-[2-(dimethylamino-κN)phenyl]-N1,N1-dimethyl-1,2-benzenediaminato-κN1,κN2]-nickel, Nickamine
About This Item
Recommended Products
Assay
≥97% (AT)
form
lumps
reaction suitability
core: nickel
reagent type: catalyst
SMILES string
CN(C)c1ccccc1N([Ni]Cl)c2ccccc2N(C)C
InChI
1S/C16H20N3.ClH.Ni/c1-18(2)15-11-7-5-9-13(15)17-14-10-6-8-12-16(14)19(3)4;;/h5-12H,1-4H3;1H;/q-1;;+2/p-1
InChI key
IRLHVNUEEKJBEL-UHFFFAOYSA-M
Related Categories
Application
- Alkyl-alkyl Kumada coupling of secondary alkyl halides
- Direct alkylation of heterocyclic C-H bonds
- Sonogashira coupling of nonactivated alkyl halides
- Kumada-Corriu-Tamao coupling of nonactivated alkyl halides with aryl and heteroaryl nucleophiles
Reactant of pincer NN2 ligand leading to selective carbon-carbon bond formation
Packaging
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service