Skip to Content
Merck
All Photos(1)

Key Documents

244996

Sigma-Aldrich

Tetrakis(triphenylphosphine)nickel(0)

Ni 4-7 % (approx.)

Synonym(s):

Ni(PPh3)4, Nickel-tetrakis(triphenylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]4Ni
CAS Number:
Molecular Weight:
1107.84
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

composition

Ni, 4-7% (approx.)

Quality Level

reaction suitability

core: nickel
reagent type: catalyst

SMILES string

[Ni].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Ni/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

KFBKRCXOTTUAFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for coupling reactions of organic halides, organometallic Compounds and also for the cyclooligomerization of cumulenes;.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Negishi, E-i.; Takahashi, et al.
Journal of the American Chemical Society, 109, 2393-2393 (1987)
Peter Werhun et al.
Inorganic chemistry, 56(16), 9996-10006 (2017-08-05)
Despite the significance of nickel compounds, NMR spectroscopy of the active nickel isotope
Iyoda, M.; Tanaka, S. et al.
Journal of the American Chemical Society, 110, 8494-8494 (1988)
Iyoda, M.; Kuwatani, Y.; Oda, M.
Journal of the American Chemical Society, 111, 3761-3761 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service