Skip to Content
Merck
All Photos(3)

Key Documents

723614

Sigma-Aldrich

4-(Diphenylamino)phenylboronic acid pinacol ester

95%

Synonym(s):

N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl-N-phenylbenzenamine, Diphenyl-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BNO2
Molecular Weight:
371.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

powder

mp

93-98 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(cc2)N(c3ccccc3)c4ccccc4

InChI

1S/C24H26BNO2/c1-23(2)24(3,4)28-25(27-23)19-15-17-22(18-16-19)26(20-11-7-5-8-12-20)21-13-9-6-10-14-21/h5-18H,1-4H3

InChI key

VKSWIFGDKIEVFZ-UHFFFAOYSA-N

General description

4-(Diphenylamino)phenylboronic acid pinacol ester is an aryl boronic acid ester that is majorly used in organic synthesis. It can be used in the transition metal-catalyzed Suzuki-Miyaura cross-coupling reaction due to its low toxicity and unique reactivity. It is an electron rich boronic acid ester that can also be used in protodeboronation.

Application

4-(Diphenylamino)phenylboronic acid pinacol ester may be used to synthesize 4-(2,2′ -bithiophen-5-yl)- 5-phenylpyrimidine for potential usage in the development of sensing devices for the detection of nitroaromatic explosives. It can also be used in the synthesis of oligothiophene (electron donating group) for the fabrication of dye sensitized solar cells (DSSCs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Base-promoted silver-catalyzed protodeboronation of arylboronic acids and esters
Liu C, et al.
Royal Society of Chemistry Advances, 5(20), 15354-15358 (2015)
New V-shaped push-pull systems based on 4, 5-di (hetero) aryl substituted pyrimidines: their synthesis and application to the detection of nitroaromatic explosives
Verbitskiy EV, et al.
ARKIVOC (Gainesville, FL, United States), 3(11), 360-373 (2016)
Triphenylamine-based dyes for dye-sensitized solar cells
Zhang F, et al.
Dyes and Pigments, 81(3), 224-230 (2009)
Fluorine in Life Sciences: Pharmaceuticals, Medicinal Diagnostics, and Agrochemicals
Progress in Fluorine Science Series (2018)
Microwave-assisted synthesis of 4-(2, 2'-bithiophen-5-yl)-5-phenylpyrimidine derivatives as sensors for detection of nitroaromatic explosives
Verbitskiy EV, et al.
Chemistry of Heterocyclic Compounds, 52(11), 904-909 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service