Skip to Content
Merck
All Photos(1)

Key Documents

711624

Sigma-Aldrich

1,3-Diiodo-5,5-dimethylhydantoin

≥96%

Synonym(s):

DIH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6I2N2O2
CAS Number:
Molecular Weight:
379.92
Beilstein:
146040
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96%

form

powder

storage temp.

−20°C

SMILES string

CC1(C)N(I)C(=O)N(I)C1=O

InChI

1S/C5H6I2N2O2/c1-5(2)3(10)8(6)4(11)9(5)7/h1-2H3

InChI key

RDZHCKRAHUPIFK-UHFFFAOYSA-N

Application

DIH - Highly efficient and economical reagent for Iodination

Used for:
  • Preparation of nitriles from corresponding alcohols and amines via oxidative conversion
  • Chemoselective, stereospecific iododesilylation of silylated alkenes
  • Preparation of N-benzyl toluenesulfonamides via sulfonylamidation of alkylbenzenes with toluenesulfonamide in presence of diiododimethylhydantoin
  • Iodination reactions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukumar Namineni et al.
Journal of hepatology, 72(5), 960-975 (2020-01-19)
Hepatic innate immune control of viral infections has largely been attributed to Kupffer cells, the liver-resident macrophages. However, hepatocytes, the parenchymal cells of the liver, also possess potent immunological functions in addition to their known metabolic functions. Owing to their
James Liang et al.
Journal of clinical pathology, 71(10), 916-925 (2018-06-03)
The number of precursor and mature lymphoid cells and plasma cells in normal bone marrow trephine (BMT) biopsies throughout the human lifespan is unknown. Reference ranges have been established from aspirated marrow, but due to haemodilution errors, these do not
AngAng Ding et al.
Journal of vascular surgery. Venous and lymphatic disorders, 7(5), 715-723 (2019-08-20)
For vascular anomalies, when clinical findings are not sufficient, auxiliary examination is essential. In this study, we characterize and differentiate the ultrasound (US) findings of deep infantile hemangioma (DIH) and venous malformation (VM). A total of 135 patients (140 lesions)
Pamela M Vacek et al.
Occupational and environmental medicine, 74(3), 211-217 (2016-11-09)
To understand why 2 studies relating crystalline silica exposure to lung-cancer mortality in Vermont granite workers yielded conflicting results. Data used in the 2 studies were linked to identify discrepancies. Mortality data and employment histories from the earlier study were
Shirin Sabouri et al.
Journal of magnetic resonance imaging : JMRI, 46(3), 861-869 (2017-01-29)
To determine the relationship between parameters measured from luminal water imaging (LWI), a new magnetic resonance imaging (MRI) T In all, 17 patients with prostate cancer were examined with a 3D multiecho spin echo sequence at 3T prior to undergoing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service