157902
1,3-Dibromo-5,5-dimethylhydantoin
98%
Synonym(s):
Dibromantin
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C5H6Br2N2O2
CAS Number:
Molecular Weight:
285.92
Beilstein:
146024
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
197-199 °C (dec.) (lit.)
SMILES string
CC1(C)N(Br)C(=O)N(Br)C1=O
InChI
1S/C5H6Br2N2O2/c1-5(2)3(10)8(6)4(11)9(5)7/h1-2H3
InChI key
VRLDVERQJMEPIF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Useful in aromatic bromination of alkoxybenzoic acids and in bromofluorination of alkenes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M Hilp
Die Pharmazie, 59(8), 612-614 (2004-09-24)
Iodine values (iodine numbers) of several fixed oils and lard can be determined in ethyl acetate, an easily biodegredable solvent, instead of chloroform according to PH. EUR. 2002. Iodine monobromide has been replaced by 1,3-dibromo-5,5-dimethylhydantoin (DBH) and potassium iodide (KI)
Soni Kamlesh Madhusudan et al.
Carbohydrate research, 340(3), 497-502 (2005-02-01)
Exchange of acyclic glycosyl dithioacetals to their O,O-acetals has been achieved by a generalized reaction protocol mediated by 1,3-dibromo-5,5-dimethylhydantoin under mild, metal-free and neutral conditions. This methodology has been extended to the synthesis of alkyl glycofuranosides.
M Hilp
Die Pharmazie, 57(6), 393-395 (2002-07-16)
Elemental bromine serves as oxidant for the identification of propylthiouracil, 2-thiouracil and sulphur according to PH. EUR. 2002. Phenol is identified according to PH. EUR. 2002 with bromine water as the sparingly water-soluble 2,4,4,6-tetrabromo-2,5-cyclohexadiene-1-one. These tests can be performed better
M Hilp
Die Pharmazie, 56(7), 548-551 (2001-08-08)
The identification of lactate according to Ph. Eur. 1997 and DAB 2000 uses the oxidation of lactic acid to pyruvic acid by boiling with bromine water in sulphuric acid. Acetaldehyde arising by decarboxylation is detected according to Legal applying a
M Hilp
Die Pharmazie, 57(1), 45-48 (2002-02-12)
PH. EUR. 2002 uses elemental bromine performing iron limit tests for maleic acid (iron 5 ppm) and titanium dioxide (iron 200 ppm). 1,3-Dibromo-5,5-dimethylhydantoin (DBH) can replace bromine water. For the iron limit test of maleic acid bivalent iron is oxidized
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service