Skip to Content
Merck
All Photos(1)

Key Documents

710717

Sigma-Aldrich

Trichlorofluoromethane solution

NMR reference standard, 10% in acetone-d6 (99.9 atom % D), TMS 6 %, 1,2-dichloro-1,1-difluoroethane 1 %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

Quality Level

form

solution

composition

1,2-dichloro-1,1-difluoroethane, 1%
TMS, 6%

concentration

10% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

SMILES string

FC(Cl)(Cl)Cl

InChI

1S/CCl3F/c2-1(3,4)5

InChI key

CYRMSUTZVYGINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Ozone 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konrad Stemmler et al.
Environmental science & technology, 41(4), 1145-1151 (2007-06-28)
HFC-365mfc (1,1,1,3,3-pentafluorobutane) is an industrial chemical used for polyurethane foam blowing. From early 2003, HFC-365mfc has been commercially produced as a substitute for HCFC-141b, whose use in Europe has been banned since January 2004. We describe the first detection of
A Vinegar
Toxicological sciences : an official journal of the Society of Toxicology, 60(1), 20-27 (2001-02-27)
Human exposure guidelines for halogenated hydrocarbons (halons) and halon replacement chemicals have been established using dose-response data obtained from canine cardiac sensitization studies. In order to provide a tool for decision makers and regulators tasked with setting guidelines for egress
A Groppi et al.
Journal of forensic sciences, 39(3), 871-876 (1994-05-01)
A case of lethal poisoning due to trichlorofluoromethane (FC11) inhalation is described. The fluorocarbon was determined in biological tissues by headspace gas chromatography-mass spectrometry. FC11 was detected in all the examined tissues, with decreasing levels in heart, lung, brain, liver
E Jüngling et al.
Analytical biochemistry, 171(1), 150-157 (1988-05-15)
A one-vial procedure has been developed to quantify free fatty acids in human blood serum and rat heart tissue. To allow routine analysis the system has been constructed to allow simultaneous processing of nine samples (Praechromat). The free fatty acids
P Sharma et al.
Rapid communications in mass spectrometry : RCM, 18(20), 2383-2387 (2004-09-24)
Nonresonant laser-induced multiphoton dissociation/ionization studies have been conducted for trichloromethane (CHCl3) and trichlorofluoromethane (CFCl3) at 355 nm, using time-of-flight mass spectrometry (TOFMS). The molecular ion signal was found to be missing for both these compounds, and very similar fragmentation patterns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service