Skip to Content
Merck
All Photos(1)

Documents

708046

Sigma-Aldrich

Ethylbenzene solution

NMR reference standard, 10% in chloroform-d (99.8 atom % D)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

concentration

10% in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.
5 mm × 8 in.

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Related Categories

Features and Benefits

1H sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murthy Kasi et al.
Biodegradation, 24(2), 279-293 (2012-08-16)
A comprehensive study on the effects of different carbon sources during the bacterial enrichment on the removal performances of benzene, toluene, ethylbenzene, and xylenes (BTEX) compounds when present as a mixture was conducted. Batch BTEX removal kinetic experiments were performed
Carla G A da Silva et al.
Journal of chromatography. A, 1260, 81-87 (2012-09-12)
Endcapped stationary phases were prepared after thermal immobilization of poly(methyloctadecylsiloxane) (PMODS) onto zirconized and titanized silica supports. These new stationary phases have lower densities of residual hydroxyl groups, according to infrared spectroscopy and 29Si CP-MAS NMR and as shown by
Adrián Flores Orozco et al.
Journal of contaminant hydrology, 136-137, 131-144 (2012-07-13)
Broadband spectral induced polarization (SIP) measurements were conducted at a former hydrogenation plant in Zeitz (NE Germany) to investigate the potential of SIP imaging to delineate areas with different BTEX (benzene, toluene, ethylbenzene, and xylene) concentrations. Conductivity images reveal a
Bundesgesundheitsblatt, Gesundheitsforschung, Gesundheitsschutz, 55(9), 1192-1200 (2012-09-01)
The German Working Group on Indoor Guidelines of the Federal Environment Agency and the States' Health Authorities is issuing indoor air guide values to protect public health. No human studies are available for health evaluation of ethylbenzene in indoor air.
Daniel Knack et al.
Applied and environmental microbiology, 78(18), 6475-6482 (2012-07-10)
Ethylbenzene dehydrogenase (EbDH) catalyzes the initial step in anaerobic degradation of ethylbenzene in denitrifying bacteria, namely, the oxygen-independent hydroxylation of ethylbenzene to (S)-1-phenylethanol. In our study we investigate the kinetic properties of 46 substrate analogs acting as substrates or inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service