Skip to Content
Merck
All Photos(1)

Key Documents

611905

Sigma-Aldrich

1,4-Dioxane solution

NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

composition

chromium(III) acetylacetonate, 5 mg/mL

concentration

40% in benzene-d6 (99.6 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

13C sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Blood, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

70.0 °F - closed cup

Flash Point(C)

21.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoyuki Kishimoto et al.
Water research, 47(5), 1919-1927 (2013-02-09)
This paper reports on the reusability and the optimal reuse of iron-rich sludge for an electrochemical Fenton-type process using sequencing batch mode and separation batch mode reuse models. In the sequencing batch mode, processes of electrochemical treatment, neutralization, sedimentation, and
Megha Aggarwal et al.
PloS one, 7(12), e51288-e51288 (2012-12-20)
The nucleocapsid core interaction with endodomains of glycoproteins plays a critical role in the alphavirus life cycle that is essential to virus budding. Recent cryo-electron microscopy (cryo-EM) studies provide structural insights into key interactions between capsid protein (CP) and trans-membrane
O N Makshakova et al.
Bioorganicheskaia khimiia, 38(3), 300-305 (2012-09-25)
Our interest in modifying influence of the low-molecular organic compounds on the hydration shell of biopolymers is due to a well-known fact that the former can act as regulators of the enzymatic activity changed the hydration shell of the latter.
Daniel Gerrity et al.
Water research, 46(19), 6257-6272 (2012-10-16)
The performance of ozonation in wastewater depends on water quality and the ability to form hydroxyl radicals (·OH) to meet disinfection or contaminant transformation objectives. Since there are no on-line methods to assess ozone and ·OH exposure in wastewater, many
Carlos E Agudelo-Morales et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(18), 4195-4201 (2012-10-24)
The quenching of pyrene and 1-methylpyrene fluorescence by nitroanilines (NAs), such as 2-, 3-, and 4-nitroaniline (2-NA, 3-NA, and 4-NA, respectively), 4-methyl-3-nitroaniline (4-M-3-NA), 2-methyl-4-nitroaniline (2-M-4-NA), and 4-methyl-3,5-dinitroaniline (4-M-3,5-DNA), are studied in toluene and 1,4-dioxane. Steady-state fluorescence data show the higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service