406074
Carbon nanotube, multi-walled
powdered cylinder cores, 20-30% MWCNT basis, O.D. × L 7-12 nm × 0.5-10 μm, avg. no. of layers, 5 ‑ 20
Synonym(s):
MWCNT, MWNT, Multiwall carbon nanotube
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
Assay
20-30% MWCNT basis
form
powder
feature
avg. no. of layers 5 ‑ 20
composition
carbon content, >99% TGA
O.D. × L
7-12 nm × 0.5-10 μm
avg. part. size
−270 mesh
<53 μm
mp
3652-3697 °C (lit.)
density
~2.1 g/mL at 25 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Multi-walled carbon nanotube core surrounded by a fused carbon shell, the remainder being multi-layer polygonal carbon nanoparticles and amorphous and graphitic carbon nanoparticles.It contains approximately 5-20 graphitic layers. Carbon nanotubes produced by arc discharge technique contain very low impurity and possesses very high crystallinity. 4
Application
Bundles of MWNTs were trapped and manipulated to form nanotube ropes using optical tweezer. MWNTs were calcined and sonicated to form carbon nanoribbons. The nanoribbons were used to fabricate array of 98 source and drain electrodes on a 300 nm SiO2/p++Si substrates.2 A MWNT probe was fabricated on Si cantilevers.
Carbon nanotube, multi-walled (MWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, biosensor, gas sensor, supercapacitor and as a filler that acts as a reinforcement to improve the mechanical property of composites.
Physical form
Approximately 5-20 graphitic layers. Contains approximately 10-40% tubes, the remainder being multi-layer polygonal carbon nanoparticles and amorphous and graphitic carbon nanoparticles.
Preparation Note
Electric Arc Discharge Method
Other Notes
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Alice Marcotte et al.
Nature materials, 19(10), 1057-1061 (2020-07-15)
Fluid and ionic transport at the nanoscale has recently demonstrated a wealth of exotic behaviours1-14. However, artificial nanofluidic devices15-18 are still far from demonstrating the advanced functionalities existing in biological systems, such as electrically and mechanically activated transport19,20. Here, we
Processing carbon nanotubes with holographic optical tweezers
Plewa J, et al.
Optics Express, 12(9), 1978-1981 (2004)
Simple and reliable method of conductive SPM
probe fabrication using carbon nanotubes
Dremov V, et al.
arXiv null
Preparation of multi-walled carbon nanotube supported TiO2 and its photocatalytic activity in the reduction of CO2 with H2O
Xia X, et al.
Carbon, 45(4), 717-721 (2007)
Mechanical behavior and microstructure of cement composites incorporating surface-treated multi-walled carbon nanotubes
Li GY, et al.
Carbon, 43(6), 1239-1245 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service