Skip to Content
Merck
All Photos(2)

Key Documents

695904

Sigma-Aldrich

1,3-Butadiene solution

15 wt. % in hexane

Synonym(s):

Bivinyl, Vinylethylene, alpha,gamma-Butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6
CAS Number:
Molecular Weight:
54.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

15 wt. % in hexane

refractive index

n20/D 1.376

density

0.682 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=CC=C

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2

InChI key

KAKZBPTYRLMSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,3-Butadiene can be used as a monomer to synthesize polybutadiene with one hydroxy end group and other silyl hydroxy-protecting end group, a precursor for two-step synthesis of hydroxyl-terminated polybutadiene (HTPB).

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-105.0 °F

Flash Point(C)

-76.11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of hydroxyl-terminated polybutadiene bearing pendant carboxyl groups by combination of anionic polymerization and blue light photocatalytic thiol-ene reaction and its pH-triggered self-assemble behavior.
Zhang W, et al.
Reactive and Functional Polymers, 127, 161-167 (2018)
Xin Min et al.
Royal Society open science, 5(5), 180156-180156 (2018-06-13)
A novel alkyl lithium-based initiator with relatively large steric hindrance, tert-butyldimethylsiloxydimethylpropyl lithium (TBDMSODPrLi), was designed and synthesized. By using TBDMSODPrLi, hydroxyl-terminated polybutadiene (HTPB) was prepared via anionic polymerization. The macromolecular structure of HTPB was characterized and verified by FTIR and
Dewakar Sangaraju et al.
Analytical chemistry, 84(3), 1732-1739 (2012-01-10)
1,3-Butadiene (BD) is an important industrial chemical and a common environmental pollutant present in urban air. BD is classified as a human carcinogen based on epidemiological evidence for an increased incidence of leukemia in workers occupationally exposed to BD and
Masato Ohashi et al.
Journal of the American Chemical Society, 133(45), 18018-18021 (2011-10-19)
Pyridines, which comprise one of the most important classes of the six-membered heterocyclic compounds, are widely distributed in nature, and the transition-metal-catalyzed [2 + 2 + 2] cycloaddition reaction of two alkynes and a nitrile is one of the most
Matthew S McCammant et al.
Journal of the American Chemical Society, 135(11), 4167-4170 (2013-03-12)
A palladium-catalyzed 1,4-addition across the commodity chemical 1,3-butadiene to afford skipped polyene products is reported. Through a palladium σ → π → σ allyl isomerization, two new carbon-carbon bonds are formed with high regioselectivity and trans stereoselectivity of the newly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service