Skip to Content
Merck
All Photos(2)

Key Documents

690872

Sigma-Aldrich

Basolite® F300

greener alternative

produced by BASF

Synonym(s):

Fe-BTC, Iron 1,3,5-benzenetricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H3FeO6
CAS Number:
Molecular Weight:
262.96
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

produced by BASF

Quality Level

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

120-190 m2/g

bulk density

0.16‑0.35 g/cm3

greener alternative category

SMILES string

O=C1O[Fe]2OC(=O)c3cc1cc(c3)C(=O)O2

InChI

1S/C9H6O6.Fe/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15;/h1-3H,(H,10,11)(H,12,13)(H,14,15);/q;+3/p-3

InChI key

UTYFBNKMZYRAFO-UHFFFAOYSA-K

General description

Basolite®F300 (Fe-BTC) is a porous metal organic framework (MOF) that is used as a highly active catalyst for Lewis acid reactions such as acetalization of aldehydes and aldol condensation. It has a hybrid supertetrahedral build up from oxocentered trimmers of iron(III) octahedron that are connected by trimesate anions.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Fe-BTC based MOF can be used in greener applications such as energy storage through lithium ion batteries. It can also be used in the supercritical adsorption of CO2.

Legal Information

Basolite is a registered trademark of BASF SE

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A combined computational and experimental study of high pressure and supercritical CO2 adsorption on Basolite MOFs
Deniz E, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 175(115), 34-42 (2013)
Vijaya Ravinayagam et al.
Saudi journal of biological sciences, 27(7), 1726-1736 (2020-06-23)
Drug resistant bacteria affects millions worldwide and remains a serious threat to health care system. The study reports the first application of hybrid nanocomposites based on zeolitic imidazolate framework-8 (ZIF-8) with MFI structured zeolite Ti-ZSM-5 (TiZ5) and mesoporous carbon (MC).
Comparison of porous iron trimesates basolite F300 and MIL-100 (Fe) as heterogeneous catalysts for lewis acid and oxidation reactions: roles of structural defects and stability
Dhakshinamoorthy A, et al.
ACS Catalysis, 2(10), 2060-2065 (2012)
Facile synthesis of the Basolite F300-like nanoscale Fe-BTC framework and its lithium storage properties
Hu X, et al.
Royal Society of Chemistry Advances, 6(115), 114483-114490 (2016)
Direct synthesis, structural features, and enhanced catalytic activity of the basolite F300-like semiamorphous Fe-BTC framework
Sanchez-Sanchez M, et al.
Crystal Growth & Design, 15(9), 4498-4506 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service