Skip to Content
Merck
All Photos(1)

Key Documents

688487

Sigma-Aldrich

2-Mesityl-2,5,6,7-tetrahydropyrrolo[2,1-c][1,2,4]triazol-4-ium chloride

97%

Synonym(s):

Bode Catalyst 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18ClN3
CAS Number:
Molecular Weight:
263.77
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

200-205 °C

SMILES string

[Cl-].Cc1cc(C)c(c(C)c1)-[n+]2cn3CCCc3n2

InChI

1S/C14H18N3.ClH/c1-10-7-11(2)14(12(3)8-10)17-9-16-6-4-5-13(16)15-17;/h7-9H,4-6H2,1-3H3;1H/q+1;/p-1

InChI key

KKBONGWVMVUYPA-UHFFFAOYSA-M

Application

Catalyst for:
  • Intermolecular hydroacylation of cyclopropenes
  • Crossed acyloin condensations
  • Domino ring-opening redox amidation Knoevenagel condensation
  • Claisen rearrangement
  • Lewis acid- and N-heterocyclic carbene-catalyzed cyclocondensation reaction
  • Bode catalyst for enantioselective cyclizations

Legal Information

Sold in collaboration with BioBlock, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service