Skip to Content
Merck
All Photos(1)

Key Documents

674788

Sigma-Aldrich

5a(R),10b(S)-5a,10b-Dihydro-2-(pentafluorophenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium tetrafluoroborate

97%

Synonym(s):

2-Pentafluorophenyl-6,10b-dihydro-4H,5aH-5-oxo-3,10c-diaza-2-azoniacyclopenta[c]fluorine tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11BF9N3O
CAS Number:
Molecular Weight:
467.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

233-237 °C

functional group

ether
fluoro

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.Fc1c(F)c(F)c(c(F)c1F)-[n+]2c[n@@H]3[C@@H]4[C@@H](Cc5ccccc45)OCc3n2

InChI

1S/C18H11F5N3O.BF4/c19-12-13(20)15(22)18(16(23)14(12)21)26-7-25-11(24-26)6-27-10-5-8-3-1-2-4-9(8)17(10)25;2-1(3,4)5/h1-4,7,10,17H,5-6H2;/q+1;-1/t10-,17+;/m1./s1

InChI key

CPCMDOOTVHDRTM-CVJFODCESA-N

Application

5a(R),10b(S)-5a,10b-Dihydro-2-(pentafluorophenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium tetrafluoroborate can be used as an N-heterocyclic carbene (NHC) organocatalyst in:
  • Benzoin reactions of aldehydes and total synthesis of a natural product named isodarparvinol B.
  • Synthesis of spirocyclic oxindole-dihydropyranones by reacting α-bromo-α,β-unsaturated aldehydes with isatin derivatives.
  • Synthesis of 1,4-dicarbonyl compounds through intramolecular Stetter reaction.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-heterocyclic carbene/Lewis acid strategy for the stereoselective synthesis of spirocyclic oxindole--dihydropyranones
Xiao Z, et al.
Organic Letters, 16(14), 3632-3635 (2014)
Scope of the asymmetric intramolecular Stetter reaction catalyzed by chiral nucleophilic triazolinylidene carbenes
de Alaniz JR, et al.
The Journal of Organic Chemistry, 73(6), 2033-2040 (2008)
Minimization of Amounts of Catalyst and Solvent in NHC-Catalyzed Benzoin Reactions of Solid Aldehydes: Mechanistic Consideration of Solid-to-Solid Conversion and Total Synthesis of Isodarparvinol B
Iwai K, et al.
ACS Omega, 5(17), 10207-10216 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service