All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
mp
29-36 °C
SMILES string
CC(C)(C)OC(=O)N1CCCCC1=O
InChI
1S/C10H17NO3/c1-10(2,3)14-9(13)11-7-5-4-6-8(11)12/h4-7H2,1-3H3
InChI key
ULMHMJAEGZPQRY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Marvin J Meyers et al.
Bioorganic & medicinal chemistry, 23(16), 5144-5150 (2015-03-24)
Given the rise of parasite resistance to all currently used antimalarial drugs, the identification of novel chemotypes with unique mechanisms of action is of paramount importance. Since Plasmodium expresses a number of aspartic proteases necessary for its survival, we have
Marius Brehm et al.
Small (Weinheim an der Bergstrasse, Germany), 16(10), e1905971-e1905971 (2020-01-28)
Miniaturization and parallelization of combinatorial organic synthesis is important to accelerate the process of drug discovery while reducing the consumption of reagents and solvents. This work presents a miniaturized platform for on-chip solid-phase combinatorial library synthesis with UV-triggered on-chip cell
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service