685879
(1S,2S)-1,2-Bis(2-hydroxyphenyl)ethylenediamine
95%
Synonym(s):
(1S,2S)-1,2-Bis(2-hydroxyphenyl)-1,2-ethanediamine, (S,S) Jik Chin Mother Diamine, (S,S) Diaminopharm Diamine, (S,S) Mother Diamine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C14H16N2O2
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
optical activity
[α]22/D -65°, c = 0.2 in chloroform
mp
157-162 °C
functional group
amine
SMILES string
N[C@H]([C@@H](N)c1ccccc1O)c2ccccc2O
InChI
1S/C14H16N2O2/c15-13(9-5-1-3-7-11(9)17)14(16)10-6-2-4-8-12(10)18/h1-8,13-14,17-18H,15-16H2/t13-,14-/m0/s1
InChI key
MRNPLGLZBUDMRE-KBPBESRZSA-N
Related Categories
General description
1,2-Bis(2-hydroxyphenyl)ethylenediamine is a chiral diamine, which acts as a key precursor (mother diamine) in the synthesis of alkyl, aryl, and heterocyclic diamines (daughter diamines), used for various synthetic applications.
Application
(1S,2S)-1,2-Bis(2-hydroxyphenyl)ethylenediamine can be used:
- As a starting material for the synthesis of Schiff base complexes of gold(III), bearing potent anticancer activity.
- As a stereoinductor in the synthesis of quinoline and isoquinoline based 1,2-diamines; that are employed as catalysts in the preparation of warfarin and coumachlor in water.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mimicking nature to make unnatural amino acids and chiral diamines
So SM, et al.
European Journal of Organic Chemistry, 2012(2), 229-241 (2012)
Green asymmetric synthesis of Warfarin and Coumachlor in pure water catalyzed by quinoline-derived 1, 2-diamines
Kucherenko AS, et al.
Green Chemistry, 20(3), 754-759 (2018)
Synthesis and biological evaluation of gold (III) Schiff base complexes for the treatment of hepatocellular carcinoma through attenuating TrxR activity
Bian M, et al.
European Journal of Medicinal Chemistry, 193(36), 112234-112234 (2020)
Stereospecific synthesis of C 2 symmetric diamines from the mother diamine by resonance-assisted hydrogen-bond directed diaza-Cope rearrangement.
Kim H, et al.
Journal of the American Chemical Society, 130(36), 12184-12191 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service