Skip to Content
Merck
All Photos(1)

Key Documents

683434

Sigma-Aldrich

(Dimethylphenylsilyl)boronic acid pinacol ester

95%

Synonym(s):

2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, B-(dimethylphenylsilyl)pinacolborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23BO2Si
CAS Number:
Molecular Weight:
262.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.4946

density

0.962 g/mL at 25 °C

SMILES string

CC1(C)OB(OC1(C)C)[Si](C)(C)c2ccccc2

InChI

1S/C14H23BO2Si/c1-13(2)14(3,4)17-15(16-13)18(5,6)12-10-8-7-9-11-12/h7-11H,1-6H3

InChI key

ARMSAQNLTKGMGM-UHFFFAOYSA-N

Application

(Dimethylphenylsilyl)boronic acid pinacol ester (Suginome′s reagent) can be used as a reagent:
  • For the selective addition of dimethylphenylsilanyl group to cyclic and acyclic unsaturated ketones, esters, acrylonitriles using a copper catalyst.
  • In the synthesis of (Z)-4-boryl-1-silyl-2-alkene derivatives by stereoselective addition of silicon-boron bond to acyclic 1,3-dienes in presence of Ni catalyst.
  • In the preparation of silyl-substituted butenoate and β-silyl-substituted acrylate derivatives from allenes and propiolate derivatives via hydrosilylation reactions using a copper catalyst.
  • In the palladium-catalyzed asymmetric silaboration of allenes and alkanes to offered corresponding β-borylallylsilanes and 2-boryl-1-silylalkanes respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective conjugate silyl additions to cyclic and acyclic unsaturated carbonyls catalyzed by Cu complexes of chiral N-heterocyclic carbenes.
Lee K S and Hoveyda A H
Journal of the American Chemical Society, 132(9), 2898-2900 (2010)
Stereoselective 1, 4-silaboration of 1, 3-dienes catalyzed by nickel complexes
Suginome M, et al.
Organic Letters, 1(10), 1567-1569 (1999)
Geminal Difunctionalization of Alkenylidene?Type Carbenoids by Using Interelement Compounds.
Hata T, et al.
Angewandte Chemie (International Edition in English), 40(4), 790-792 (2001)
Kang-Sang Lee et al.
Journal of the American Chemical Society, 132(9), 2898-2900 (2010-02-13)
An efficient Cu-catalyzed protocol for enantioselective addition of a dimethylphenylsilanyl group to a wide range of cyclic and acyclic unsaturated ketones, esters, acrylonitriles, and alpha,beta,gamma,delta-dienones is disclosed. Reactions are performed in the presence of 1-2 mol % of commercially available
Platinum?Catalyzed Regioselective Silaboration of Alkenes.
Suginome M, et al.
Angewandte Chemie (International Edition in English), 36(22), 2516-2518 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service