Skip to Content
Merck
All Photos(2)

Key Documents

683132

Sigma-Aldrich

Chiralyst P493

Umicore, 98%

Synonym(s):

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Di(μ-chloro)bis(1,5-cyclooctadiene)dirhodium(I), 1,5-Cyclooctadienerhodium(I) chloride dimer, Bis(1,5-cyclooctadiene)dirhodium(I) dichloride, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]dirhodium, Rhodium(I) chloride 1,5-Cyclooctadiene complex dimer, [Rh(COD)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24Cl2Rh2
CAS Number:
Molecular Weight:
493.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

reaction suitability

core: rhodium
reaction type: Cross Couplings
reagent type: catalyst

mp

216.3-219.2 °C
243 °C (dec.) (lit.)

SMILES string

Cl[Rh].Cl[Rh].C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.2ClH.2Rh/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;

InChI key

QSUDXYGZLAJAQU-MIXQCLKLSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chloro(1,5-cyclooctadiene)rhodium(I) dimer ([Rh(COD)Cl]2) can be:
  • Employed for the synthesis of rhodium complex of heterocyclic carbenes (NHCs).
  • Modified and coated on the surface of ferrite magnetic nanoparticles for catalyzing hydroformylation reaction of olefins.
  • Used for the synthesis of triple-layer structure to be used as a ring-opening polymerization catalyst.
  • As an effective catalyst for dehydrogenation of amine-borane adducts (hydrogen storage materials) such as ammonia-borane.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allosteric supramolecular triple-layer catalysts.
Yoon HJ, et al.
Science, 330(6000), 66-69 (2010)
Rhodium and iridium complexes of N-heterocyclic carbenes via transmetalation: structure and dynamics.
Chianese AR, et al.
Organometallics, 22(8), 1663-1667 (2003)
Highly Efficient Colloidal Cobalt-and Rhodium-Catalyzed Hydrolysis of H3N?BH3 in Air.
Clark TJ, et al.
Inorganic Chemistry, 46(18), 7522-7527 (2007)
Magnetic nanoparticles as a catalyst vehicle for simple and easy recycling.
Yoon TJ, et al.
New. J. Chem., 27(2), 227-229 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service