Skip to Content
Merck
All Photos(1)

Key Documents

682322

Sigma-Aldrich

(R)-N-Methyl-N-diphenylphosphino-1-[(S)-2-diphenylphosphino)ferrocenyl]ethylamine

≥96%

Synonym(s):

(1R)-1-(Diphenylphosphino)-2-[(1R)-1-[(diphenylphosphino)methylamino]ethyl]ferrocene, Methyl-BoPhoz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H35FeNP2
CAS Number:
Molecular Weight:
611.47
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

form

solid

mp

108.7-113.6 °C

functional group

amine
phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)N(C)P(c5ccccc5)c6ccccc6

InChI

1S/C32H30NP2.C5H5.Fe/c1-26(33(2)35(29-20-11-5-12-21-29)30-22-13-6-14-23-30)31-24-15-25-32(31)34(27-16-7-3-8-17-27)28-18-9-4-10-19-28;1-2-4-5-3-1;/h3-26H,1-2H3;1-5H;/t26-;;/m1../s1

InChI key

ZIAZHUWEJYUGEO-FBHGDYMESA-N

Application

(R)-N-Methyl-N-diphenylphosphino-1-[(S)-2-diphenylphosphino)ferrocenyl]ethylamine-rhodium complex can efficiently catalyze the asymmetric hydrogenation of 2-tert-butoxycarbonylamino-3-cyclopropylacrylic acid benzyl ester to form (S)-2-tert-butoxycarbonylamino-3-cyclopropylpropionic acid benzyl ester.
Efficient ligand for asymmetric hydrogenation

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US6590115 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of enantiomerically pure cyclopropylalanine.
Boaz NW, et al.
Tetrahedron Asymmetry, 14(22), 3575-3580 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service