Skip to Content
Merck
All Photos(1)

Key Documents

379344

Sigma-Aldrich

4,5-Diazafluoren-9-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6N2O
CAS Number:
Molecular Weight:
182.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

214-217 °C (lit.)

solubility

THF: soluble(lit.)
benzene: soluble(lit.)
dichloromethane: soluble(lit.)
toluene: soluble(lit.)

functional group

ketone

SMILES string

O=C1c2cccnc2-c3ncccc13

InChI

1S/C11H6N2O/c14-11-7-3-1-5-12-9(7)10-8(11)4-2-6-13-10/h1-6H

InChI key

PFMTUGNLBQSHQC-UHFFFAOYSA-N

General description

4,5-Diazafluoren-9-one is a heterocyclic building block used in the synthesis of various heterocyclic compounds and also as a fluorescent probe.

Application

4,5-Diazafluoren-9-one may be used in the preparation of heterocyclic ligands, by condensation with various diamines, hydrazine, 1,4-phenylenediamine, benzidine and 4,4′-methylenedianiline. It may be used in the preparation of 9-diazo-4,5-diazafluorene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4,5-Diazafluoren-9-one.
White PB and Stahl SS.
e-EROS Encyclopedia of Reagents for Organic Synthesis., 1-3 (2014)
Synthesis and Application of New Ru (II) Complexes for Dye-Sensitized Nanocrystalline TiO2 Solar Cells.
Seok WK, et al.
Bull. Korean Chem. Soc., 28(8), 1311-1311 (2011)
4, 5-Diazafluoren-9-one.
Fun H-K, et al.
Acta Crystallographica Section C, Structural Chemistry, 51(10), 2076-2078 (1995)
Synthesis, characterization, electrochemical and spectroscopic studies of two new heteroleptic Ru (II) polypyridyl complexes.
Ocakoglu K, et al.
Dyes and Pigments, 75(2), 385-394 (2007)
4, 5-Diazafluoren-9-one from the oxidation of 1, 10-phenanthroline by permanganate.
Eckhard IF and Summers LA.
Australian Journal of Chemistry, 26(12), 2727-2728 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service