Skip to Content
Merck
All Photos(2)

Key Documents

677345

Sigma-Aldrich

1,2,4-Trimethylcyclohexane, mixture of isomers

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9(CH3)3
CAS Number:
Molecular Weight:
126.24
Beilstein:
1900477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.433 (lit.)

bp

141-143 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(C)C(C)C1

InChI

1S/C9H18/c1-7-4-5-8(2)9(3)6-7/h7-9H,4-6H2,1-3H3

InChI key

VCJPCEVERINRSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Vyrides et al.
Journal of hazardous materials, 177(1-3), 836-841 (2010-01-20)
Powdered activated carbon (PAC) was added to an effluent from a submerged anaerobic membrane bioreactor (SAMBR) treating saline wastewater as a post-treatment method. The adsorption of contaminants was carried out and key Freundlich isotherm parameters were evaluated. The results showed
Oddvar Myhre et al.
Toxicological sciences : an official journal of the Society of Toxicology, 80(2), 296-303 (2004-05-14)
Previously, we have shown that exposure of cultured rat cerebellar granule cells to the hydrocarbon solvent 1,2,4-trimethylcyclohexane leads to formation of reactive oxygen species (ROS). However, the cellular mechanisms responsible for formation of ROS in these cells after exposure to
I Eide et al.
Archives of toxicology, 70(7), 397-404 (1996-01-01)
The paper describes experimental and statistical methods for toxicokinetic evaluation of mixtures in inhalation experiments. Synthetic mixtures of three C9 n-paraffinic, naphthenic and aromatic hydrocarbons (n-nonane, trimethylcyclohexane and trimethylbenzene, respectively) were studied in the rat after inhalation for 12h. The
K Zahlsen et al.
Pharmacology & toxicology, 67(5), 436-440 (1990-11-01)
The concentrations of the C9 hydrocarbons n-nonane, 1,2,4-trimethylbenzene and 1,2,4-trimethylcyclohexane were measured in rat blood, brain and perirenal fat after exposures to 1000 p.p.m. of the individual compounds. Measurements were made by head space gas chromatography at the end of
Effects of the solvent 1,2,4-trimethylcyclohexane on respiratory burst in human neutrophil granulocytes. A chemiluminescence and electron paramagnetic resonance spectrometry study.
O Myhre et al.
Annals of the New York Academy of Sciences, 893, 358-361 (2000-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service