Skip to Content
Merck
All Photos(1)

Key Documents

674281

Sigma-Aldrich

1,11-Undecanedithiol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2(CH2)9CH2SH
CAS Number:
Molecular Weight:
220.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.4983

density

0.9333 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

SCCCCCCCCCCCS

InChI

1S/C11H24S2/c12-10-8-6-4-2-1-3-5-7-9-11-13/h12-13H,1-11H2

InChI key

ODMTYGIDMVZUER-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - open cup

Flash Point(C)

> 110 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuoti Xie et al.
ACS nano, 11(1), 569-578 (2016-12-10)
The transport properties of molecular junctions based on alkanedithiols with three different methylene chain lengths were compared with junctions based on similar chains wherein every third -CH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service