Skip to Content
Merck
All Photos(1)

Key Documents

663190

Sigma-Aldrich

Ethyl 5-methylindole-2-carboxylate

97%

Synonym(s):

5-Methylindole-2-carboxylic acid ethyl ester, NSC 30928

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

160-164 °C

functional group

ester

SMILES string

CCOC(=O)c1cc2cc(C)ccc2[nH]1

InChI

1S/C12H13NO2/c1-3-15-12(14)11-7-9-6-8(2)4-5-10(9)13-11/h4-7,13H,3H2,1-2H3

InChI key

KMVFKXFOPNKHEM-UHFFFAOYSA-N

General description

Ethyl 5-methylindole-2-carboxylate (5-Methylindole-2-carboxylic acid ethyl ester) is an indole derivative. Indole ring system is an important building block or intermediate in the synthesis of many pharmaceutical agents. It is formed during the Fischer indolization of ethyl pyruvate 2-[2-(methanesulfonyloxy)-4-methyl]phenylhydrazine.

Application

  • Reactant for synthesis of oxazino[4,3-a]indoles via cascade addition-cyclization reactions
  • Reactant for preparation of indolecarboxamides as cannabinoid CB1 receptor antagonists
  • Reactant for preparation of indole-3-propionic acids as antiinflammatory and analgesic agents
  • Reactant for Friedel-Crafts acylation with nitrobenzoyl chloride
  • Reactant for oximation reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substituent Effects in the Fischer Indolization of (2-Sulfonyloxyphenyl) hydrazones (Fischer Indolization and Related Compounds. XXX).
Murakami Y, et al.
Chemical & Pharmaceutical Bulletin, 47, 791-797 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service