Skip to Content
Merck
All Photos(2)

Key Documents

663042

Sigma-Aldrich

CX22

Umicore, 97%

Synonym(s):

Allylchloro[1,3-bis-(2,4,6-trimethylphenyl)imidazol-2-ylidene]palladium(II), Allyl[1,3-bis(mesityl)imidazol-2-ylidene]chloropalladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29ClN2Pd
CAS Number:
Molecular Weight:
487.37
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

228-232 °C (dec.)

SMILES string

CC=C.Cc1cc(C)c(N2C=CN(C2[Pd]Cl)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H25N2.C3H6.ClH.Pd/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-3-2;;/h7-13H,1-6H3;3H,1H2,2H3;1H;/q;;;+1/p-1

InChI key

UZZBPJRGCGHWDS-UHFFFAOYSA-M

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service