Skip to Content
Merck
All Photos(2)

Documents

660272

Sigma-Aldrich

CX12

Umicore

Synonym(s):

1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene (1,4-naphthoquinone)palladium(0) dimer, Bis[1,3-dihydro-1,3-(2,4,6-trimethylphenyl)-2H-imidazol-2-ylidene]bis[μ-[(2,3-η)-1,4-naphthalenedione-κO]]di-palladium, Naphthoquinone-1,3-bis(2,4,6-trimethylphenyl)-imidazole-2-ylidenepalladium(0), Pd(0)-naphthochinone-1,3-Bis(2,4,6-tri-methylphenyl)imidazol-2-ylidene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H60N4O4Pd2
CAS Number:
Molecular Weight:
1138.00
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

description

Umicore

Quality Level

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

>300 °C

SMILES string

O=C1C=CC(=O)c2ccccc12.O=C3C=CC(=O)c4ccccc34.Cc5cc(C)c(N6C=CN(C6=[Pd])c7c(C)cc(C)cc7C)c(C)c5.Cc8cc(C)c(N9C=CN(C9=[Pd])c%10c(C)cc(C)cc%10C)c(C)c8

InChI

1S/2C21H24N2.2C10H6O2.2Pd/c2*1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;2*11-9-5-6-10(12)8-4-2-1-3-7(8)9;;/h2*7-12H,1-6H3;2*1-6H;;

InChI key

ZTBFODSKEBBIJJ-UHFFFAOYSA-N

General description

1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene (1,4-naphthoquinone)palladium(0) dimer is an N-heterocyclic carbene based palladium catalyst that shows high catalytic activity in C-C coupling reactions.

Application

It can be used as a catalyst in the Kumada cross-coupling of aryl magnesium reagents with functionalized alkyl chlorides at room temperature to form substituted alkanes.

Other Notes

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysts for Cross-Coupling Reactions with Non-activated Alkyl Halides.
Frisch AC and Beller M.
Angewandte Chemie (International Edition in English), 44(5), 674-688 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service