Skip to Content
Merck
All Photos(1)

Key Documents

653071

Sigma-Aldrich

Ethylenebis(chloroformate)

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2CH2CH2O2CCl
CAS Number:
Molecular Weight:
186.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.436 (lit.)
n20/D 1.4500

bp

53-55 °C/0.3 mmHg (lit.)
53-55 °C/0.3 mmHg

density

1.436 g/mL at 25 °C (lit.)
1.4364 g/mL at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(=O)OCCOC(Cl)=O

InChI

1S/C4H4Cl2O4/c5-3(7)9-1-2-10-4(6)8/h1-2H2

InChI key

FMACOGSACRRIMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Qiang Wang et al.
International journal of nanomedicine, 7, 693-704 (2012-02-24)
Polyethylenimine (PEI), especially PEI 25 kDa, has been widely studied for delivery of nucleic acid drugs both in vitro and in vivo. However, it lacks degradable linkages and is too toxic for therapeutic applications. Hence, low-molecular-weight PEI has been explored

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service