Skip to Content
Merck
All Photos(2)

Documents

652482

Sigma-Aldrich

Carbon nanotube, single-walled

octadecylamine functionalized, 80-90% carbon basis, D× L 2-10 nm × 0.5-2 μm , bundle dimensions

Synonym(s):

Carbon nanotube, single-walled, ODA functionalized, SWNT, ODA functionalized

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Carbon nanotube, single-walled, octadecylamine functionalized, 80-90% carbon basis, D × L 2-10 nm × 0.5-2 μm , bundle dimensions, avg. no. of layers, 1

Assay

80-90% carbon basis

form

powder

feature

avg. no. of layers 1

extent of labeling

30-40 wt. % (ODA)

D × L

2-10 nm × 0.5-2 μm , bundle dimensions

impurities

2-6% metals

functional group

amine

General description

The average diameter of the octadecylamine functionalized single walled carbon nanotubes (ODA functionalized SWNTs) is 1.4nm ± 0.1nm. This single walled carbon nanotube (SWNT) exhibits properties normally associated with molecular species such as solubility in organic solvents and solution-based chemical transformations. The nanotubes were prepared by electric arc discharge method.

Packaging

Packaged in glass bottles

Preparation Note

Electric Arc Discharge Method

Analysis Note

Carbonaceous content

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution properties of single-walled carbon nanotubes
Chen J, et al.
Science, 282, 95-98 (1998)
S Niyogi et al.
Accounts of chemical research, 35(12), 1105-1113 (2002-12-18)
In this Account we highlight the experimental evidence in favor of our view that carbon nanotubes should be considered as a new macromolecular form of carbon with unique properties and with great potential for practical applications. We show that carbon
Nano Letters, 3, 309-309 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service