Skip to Content
Merck
All Photos(1)

Key Documents

652024

Sigma-Aldrich

4-Hydroxyphenethyl bromide

96%

Synonym(s):

2-(4-Hydroxyphenyl)-1-bromoethane, 4-(2-Bromoethyl)phenol, 4-Hydroxy-1-(2-bromoethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrO
CAS Number:
Molecular Weight:
201.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%

Quality Level

Assay

96%

form

solid

mp

88-92 °C (lit.)

functional group

bromo

SMILES string

Oc1ccc(CCBr)cc1

InChI

1S/C8H9BrO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6H2

InChI key

DYYVTFCYVZEQDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Hammerl et al.
Journal of agricultural and food chemistry, 65(15), 3230-3241 (2017-04-07)
A novel differential off-line LC-NMR approach (DOLC-NMR) was developed to capture and quantify nutrient-induced metabolome alterations in Saccharomyces cerevisiae. Off-line coupling of HPLC separation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service