Skip to Content
Merck
All Photos(1)

Key Documents

156434

Sigma-Aldrich

Cyclobutanol

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7OH
CAS Number:
Molecular Weight:
72.11
Beilstein:
2035937
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

123 °C/733 mmHg (lit.)

density

0.921 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OC1CCC1

InChI

1S/C4H8O/c5-4-2-1-3-4/h4-5H,1-3H2

InChI key

KTHXBEHDVMTNOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service