Skip to Content
Merck
All Photos(3)

Key Documents

637386

Sigma-Aldrich

6-Chloro-3-pyridinylboronic acid

≥95.0%

Synonym(s):

2-Chloro-5-pyridineboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BClNO2
CAS Number:
Molecular Weight:
157.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

mp

165 °C (lit.)

functional group

chloro

SMILES string

OB(O)c1ccc(Cl)nc1

InChI

1S/C5H5BClNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3,9-10H

InChI key

WPAPNCXMYWRTTL-UHFFFAOYSA-N

Application

6-Chloro-3-pyridinylboronic acid can be used:
  • To prepare biologically significant 3-arylcoumarins by reacting with 3-chlorocoumarin through Suzuki reaction.
  • As a substrate in the synthesis of 11-(pyridinylphenyl)steroid with progesterone agonist/antagonist profile.
  • As a substrate in the preparation of α- secondary and tertiary pyridines by the reaction of pyridotriazoles with boronic acids.
  • As a substrate in the palladium-catalyzed α-arylation of saturated cyclic amines and N-methyl amines.

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

11-(Pyridinylphenyl) steroids-A new class of mixed-profile progesterone agonists/antagonists
Rewinkel J, et al.
Bioorganic & Medicinal Chemistry, 16(6), 2753-2763 (2008)
Synthesis of 3-arylcoumarins via Suzuki-cross-coupling reactions of 3-chlorocoumarin
Matos MJ, et al.
Tetrahedron Letters, 52(11), 1225-1227 (2011)
Metal-Free Denitrogenative C-C Couplings of Pyridotriazoles with Boronic Acids To Afford α-Secondary and α-Tertiary Pyridines
Dong C, et al.
Organic Letters, 21(11), 4148-4152 (2019)
α-Arylation of Saturated Azacycles and N-Methylamines via Palladium (II)-catalyzed C (sp3)-H Coupling
Spangler JE, et al.
Journal of the American Chemical Society, 137(37), 11876-11879 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service