Skip to Content
Merck
All Photos(3)

Key Documents

637173

Sigma-Aldrich

Antimony(III) oxide

nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis

Synonym(s):

Diantimony trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2O3
CAS Number:
Molecular Weight:
291.52
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38080202
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: antimony

surface area

15.6 m2/g , typical

particle size

<250 nm (TEM)

bp

1550 °C (lit.)

mp

655 °C (lit.)

bulk density

0.5‑0.6 g/mL

SMILES string

O=[Sb]O[Sb]=O

InChI

1S/3O.2Sb

InChI key

ADCOVFLJGNWWNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nona Merry M Mitan et al.
Chemosphere, 72(7), 1073-1079 (2008-05-24)
The controlled pyrolysis of polyethylene/polypropylene/polystyrene mixed with brominated high-impact polystyrene containing decabromodiphenyl ether as a brominated flame-retardant with antimony trioxide as a synergist was performed. The effect of decabromodiphenyl ether and antimony trioxide on the formation of its congeners and
Lisa Bregoli et al.
Toxicology, 262(2), 121-129 (2009-06-02)
Nanoparticles (NPs) are materials with one dimension in the range of 1-100 nm. The toxicity of NPs remains widely unknown and still poses concerns, due to the peculiar characteristics of materials in the nano-size range. We analyze the toxicity of
Koen Oorts et al.
Environmental science & technology, 42(12), 4378-4383 (2008-07-09)
Antimony trioxide (Sb2O3) is a widely used chemical that can be emitted to soil. The fate and toxicity of this poorly soluble compound in soil is insufficiently known. A silt-loam soil (pH 7.0, background 0.005 mmol Sb kg(-1)) was amended
David Kirkland et al.
Mutation research, 627(2), 119-128 (2006-12-19)
Antimony trioxide (Sb2O3, CAS 1309-64-4) is widely used as a flame retardant synergist in a number of household products, as a fining agent in glass manufacture, and as a catalyst in the manufacture of various types of polyester plastics. It
P N Gates et al.
The Science of the total environment, 205(2-3), 215-221 (1997-12-31)
No evidence could be found for the production, in culture, of methylated antimony compounds from water-insoluble or soluble antimony derivatives by the aerobes, Scopulariopsis brevicaulis or Bacillus sp. or by anaerobes associated with cot mattress materials. The study does not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service