Skip to Content
Merck
All Photos(1)

Key Documents

612073

Sigma-Aldrich

1,4-Dioxane solution

NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.11

grade

NMR reference standard

Quality Level

composition

chromium(III) acetylacetonate, 5 mg/mL

concentration

40% in benzene-d6 (99.6 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

13C sensitivity

Quantity

3 mm O.D. tube contains 0.235 mL.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Blood, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Agudelo-Morales et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(18), 4195-4201 (2012-10-24)
The quenching of pyrene and 1-methylpyrene fluorescence by nitroanilines (NAs), such as 2-, 3-, and 4-nitroaniline (2-NA, 3-NA, and 4-NA, respectively), 4-methyl-3-nitroaniline (4-M-3-NA), 2-methyl-4-nitroaniline (2-M-4-NA), and 4-methyl-3,5-dinitroaniline (4-M-3,5-DNA), are studied in toluene and 1,4-dioxane. Steady-state fluorescence data show the higher
Kiyoaki Ishimoto et al.
Biomacromolecules, 13(11), 3757-3768 (2012-09-19)
For developing broader application of biobased polymers, graft copolymers and comb polymers having poly(lactic acid) (PLA) side chains have been synthesized by using a macromonomer technique. PLA macromonomers (MMm) having a methacryloyl polymerizable group with different PLA chain length with
R Dario Arrua et al.
Journal of chromatography. A, 1273, 26-33 (2013-01-01)
Rigid monolithic cryostructures were prepared in capillary format at sub-zero temperatures and used successfully in the separation of proteins by hydrophobic interaction chromatography (HIC). The polymerization mixture consisted of poly(ethyleneglycol) diacrylate (PEGDA) M(n)∼258 as the single monomer, a mixture of
B Thanuja et al.
Ultrasonics sonochemistry, 19(6), 1213-1220 (2012-04-14)
Density (ρ), ultrasonic velocity (U), for the binary mixtures of 4-methoxy benzoin (4MB) with ethanol, chloroform, acetonitrile, benzene, and di-oxane were measured at 298K. The solute-solvent interactions and the effect of the polarity of the solvent on the type of
Sheau-Yun Dora Chiang et al.
Journal of environmental monitoring : JEM, 14(9), 2317-2326 (2012-07-25)
An intrinsic biodegradation study involving the design and implementation of innovative environmental diagnostic tools was conducted to evaluate whether monitored natural attenuation (MNA) could be considered as part of the remedial strategy to treat an aerobic aquifer contaminated with 1,4-dioxane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service