Skip to Content
Merck
All Photos(1)

Key Documents

606871

Sigma-Aldrich

ISOGRO®-15N Powder -Growth Medium

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.12

isotopic purity

98 atom % 15N

technique(s)

bio NMR: suitable
protein expression: suitable

storage temp.

−20°C

Related Categories

Application

ISOGRO®-15N Powder -Growth Medium has been used:
  • for the production of 15N histone H2A for nuclear magnetic resonance (NMR) spectroscopy studies.
  • for the production of 15N-enriched protein kinase C and H3 histone for NMR studies.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

ISOGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of ubiquitin-nucleosome recognition and regulation of 53BP1 chromatin recruitment by RNF168/169 and RAD18
Hu Q, et al.
Molecular Cell, 66(4), 473-487 (2017)
Expression and purification of the N-terminal regulatory domain of protein kinase C for biophysical studies
Cole TR and Igumenova TI
Protein Expression and Purification, 110(4), 14-21 (2015)
Sung Wook Park et al.
Scientific reports, 9(1), 10929-10929 (2019-07-31)
The rapidly accelerated fibrosarcoma (Raf) kinase is canonically activated by growth factors that regulate multiple cellular processes. In this kinase cascade Raf activation ultimately results in extracellular regulated kinase 1/2 (Erk1/2) activation, which requires Ras binding to the Ras binding
Nicholas C Fitzkee et al.
The Journal of biological chemistry, 285(23), 18072-18084 (2010-04-07)
The human immunodeficiency virus type 1 (HIV-1) integrase (IN) is a critical enzyme involved in infection. It catalyzes two reactions to integrate the viral cDNA into the host genome, 3' processing and strand transfer, but the dynamic behavior of the
Ara Celi DiCostanzo et al.
The Journal of biological chemistry, 287(33), 27997-28006 (2012-06-29)
Light chain amyloidosis is an incurable protein misfolding disease where monoclonal immunoglobulin light chains misfold and deposit as amyloid fibrils, causing organ failure and death. Previously, we determined that amyloidogenic light chains AL-09 and AL-103 do not form fibrils at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service