Skip to Content
Merck
All Photos(3)

Key Documents

552151

Sigma-Aldrich

D-Glucose-13C6,1,2,3,4,5,6,6-d7

97 atom % D, 99 atom % 13C

Synonym(s):

Deuterated Glucose, Labeled Glucose, D-Glucose-13C6,C-d7, Dextrose-13C6,C-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H5D7O6
CAS Number:
Molecular Weight:
193.15
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
97 atom % D

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

technique(s)

bio NMR: suitable
protein expression: suitable

mp

150-152 °C (lit.)

mass shift

M+13

storage temp.

room temp

SMILES string

[2H][13C](=O)[13C@]([2H])(O)[13C@@]([2H])(O)[13C@]([2H])(O)[13C@]([2H])(O)[13C]([2H])([2H])O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1/i1+1D,2+1D2,3+1D,4+1D,5+1D,6+1D

InChI key

GZCGUPFRVQAUEE-VVZLUFAVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D- Glucose is a monosaccharide (simple sugar). It is a vital source of energy for living organisms. It exists in a cyclic form, containing six carbon atoms. It is present naturally in all plants and fruits in free or bound state. It is synthesized by the process, gluconeogenesis, in the liver and kidney.

Biochem/physiol Actions

D-Glucose is involved in the metabolic processes, such as glycolysis and Kreb′s cycle for the generation of adenosine triphosphates. It is stored in the form of glycogen in liver. D-Glucose moieties constitutes to form cellulose by β-1,4-glycosidic linkage.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis of cellulose to glucose by solid acid catalysts
Huang YB and Fu Y
Green Chemistry, 15(5), 1095-1111 (2013)
Carl Öster et al.
Science advances, 5(7), eaaw6756-eaaw6756 (2019-08-09)
Ion conduction through potassium channels is a fundamental process of life. On the basis of crystallographic data, it was originally proposed that potassium ions and water molecules are transported through the selectivity filter in an alternating arrangement, suggesting a "water-mediated"
Shilpa Nargund et al.
Biotechnology progress, 31(5), 1179-1186 (2015-06-23)
(13)C-metabolic flux analysis was used to understand copper deficiency-related restructuring of energy metabolism, which leads to excessive lactate production in recombinant protein-producing CHO cells. Stationary-phase labeling experiments with U-(13)C glucose were conducted on CHO cells grown under high and limiting
Chaevien S Clendinen et al.
Analytical chemistry, 87(11), 5698-5706 (2015-05-02)
The many advantages of (13)C NMR are often overshadowed by its intrinsically low sensitivity. Given that carbon makes up the backbone of most biologically relevant molecules, (13)C NMR offers a straightforward measurement of these compounds. Two-dimensional (13)C-(13)C correlation experiments like
Julia Peukes et al.
Nature, 587(7834), 495-498 (2020-09-11)
Influenza A virus causes millions of severe cases of disease during annual epidemics. The most abundant protein in influenza virions is matrix protein 1 (M1), which mediates virus assembly by forming an endoskeleton beneath the virus membrane1. The structure of full-length M1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service