Skip to Content
Merck
All Photos(1)

Documents

597031

Sigma-Aldrich

Bromoacetic acid-18O2

95 atom % 18O, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C18O2H
CAS Number:
Molecular Weight:
142.95
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

95 atom % 18O

Assay

97% (CP)

form

solid

bp

208 °C (lit.)

mp

47-49 °C (lit.)

mass shift

M+4

SMILES string

[18OH]C(=[18O])CBr

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i4+2,5+2

InChI key

KDPAWGWELVVRCH-MABBKULESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service