Skip to Content
Merck
All Photos(1)

Documents

602124

Sigma-Aldrich

Xenon-128Xe

25 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
128Xe
CAS Number:
Molecular Weight:
127.90
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:

isotopic purity

25 atom %

Assay

≥99.5% (Total Xenon)

form

gas

impurities

<0.5% Total others

mass shift

M-4

SMILES string

[128Xe]

InChI

1S/Xe/i1-3

InChI key

FHNFHKCVQCLJFQ-OIOBTWANSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service