Skip to Content
Merck
All Photos(1)

Key Documents

569356

Sigma-Aldrich

9,9-Dioctylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester

97%

Synonym(s):

9,9-Dioctylfluorene-2,7-bis(trimethylborate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H6O2BC6H3C(C8H17)2C6H3BO2C3H6
CAS Number:
Molecular Weight:
558.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

mp

126-130 °C (lit.)

SMILES string

CCCCCCCCC1(CCCCCCCC)c2cc(ccc2-c3ccc(cc13)B4OCCCO4)B5OCCCO5

InChI

1S/C35H52B2O4/c1-3-5-7-9-11-13-21-35(22-14-12-10-8-6-4-2)33-27-29(36-38-23-15-24-39-36)17-19-31(33)32-20-18-30(28-34(32)35)37-40-25-16-26-41-37/h17-20,27-28H,3-16,21-26H2,1-2H3

InChI key

KAYXDWIILRESPY-UHFFFAOYSA-N

General description

It′s an aryl boronate ester. Arylboronic acids may be prepared by hydrolysis of boronate esters, and consequently many other boronic acid derivatives (e.g., organotrifluoroborates) are derived.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved system for the palladium catalyzed borylation of aryl halides with pinacol borane
Billingsley KL, Buchwald SL
The Journal of Organic Chemistry, 73(14), 5589-5591 null
Paladium -catalyzed, direct boronic acide synthesis from aryl chlorides: A simplified route to diverse boronate ester derivatives
Molander GA,Trice SLJ, Dreher SD
Journal of the American Chemical Society, 132(50), 17701-17703 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service