Skip to Content
Merck
All Photos(1)

Key Documents

567752

Sigma-Aldrich

Furfuryl isocyanate

97%

Synonym(s):

(2-Furyl)methyl isocyanate, 2-(Isocyanatomethyl)furan, 2-Furanylmethyl isocyanate, 2-Furfuryl isocyanate, 2-Furfuryl isocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4760 (lit.)

bp

157-158 °C (lit.)

density

1.148 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

storage temp.

2-8°C

SMILES string

O=C=NCc1ccco1

InChI

1S/C6H5NO2/c8-5-7-4-6-2-1-3-9-6/h1-3H,4H2

InChI key

UIADMYLYGJYUSQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F - closed cup

Flash Point(C)

54.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kizkitza González et al.
International journal of biological macromolecules, 143, 265-272 (2019-12-10)
Starch-based nanocomposite hydrogels were successfully prepared by the Diels-Alder click cross-linking reaction between furan-functionalized starch derivative and a water-soluble tetrafunctional maleimide compound, adding cellulose nanocrystals (CNC) as nanoreinforcement. The effect of increasing the CNC content on rheological and swelling properties
Antoniya Toncheva et al.
ACS applied materials & interfaces, 10(35), 29933-29942 (2018-08-11)
In recent years, shape-memory polymers (SMPs) have gained a key position in the realm of actuating applications from daily life products to biomedical and aeronautic devices. Most of these SMPs rely mainly on shape changes upon direct heat exposure or
Dorothee Ehrhardt et al.
Polymers, 12(11) (2020-11-05)
Two reversible polymer networks, based on Diels-Alder cycloadditions, are selected to discuss the opportunities of mobility-controlled self-healing in ambient conditions for which information is lacking in literature. The main methods for this study are (modulated temperature) differential scanning calorimetry, microcalorimetry
Shin-Woong Kim et al.
International journal of biological macromolecules, 133, 875-880 (2019-04-29)
Systems for delivering damaged tissue by immobilization of a bioactive substance or a protein drug for rapid recovery of a patient are being studied. To immobilize drugs using natural polymer, photo-immobilization method has been designed. Immobilization through photo-reaction is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service