Skip to Content
Merck
All Photos(1)

Documents

49667

Sigma-Aldrich

4-(Maleinimido)phenyl isocyanate

purum, ≥97.0% (CHN)

Synonym(s):

N-(4-Isocyanatophenyl)maleimide, PMPI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6N2O3
CAS Number:
Molecular Weight:
214.18
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

grade

purum

Assay

≥97.0% (CHN)

form

solid

reaction suitability

reagent type: linker

mp

120-125 °C

functional group

maleimide

storage temp.

2-8°C

SMILES string

O=C(C=C1)N(C(C=C2)=CC=C2N=C=O)C1=O

InChI

1S/C11H6N2O3/c14-7-12-8-1-3-9(4-2-8)13-10(15)5-6-11(13)16/h1-6H

InChI key

OJQSISYVGFJJBY-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

A novel heterobifunctional linker for hydroxyl-to-thiol coupling

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane G Lévesque et al.
Bioconjugate chemistry, 18(3), 874-885 (2007-04-04)
Hydrogels derived from synthetic polymers have been previously engineered to degrade under the activity of matrix metalloproteinases (MMPs). It is believed that these systems can act as extracellular-matrix (ECM) equivalents mimicking the degradation and remodeling of the ECM through the
Nileshi Saraf et al.
ACS omega, 4(1), 2234-2240 (2019-02-08)
A polydimethylsiloxane-based microfluidic device has been developed for the multiplex detection of viral envelope proteins such as Zika and chikungunya on a single platform using aptamer-analyte interactions. The channel is integrated with microsized pillars that increase the surface area allowing
Xiang Chen et al.
Clinical laboratory, 66(3) (2020-03-13)
Vitamin B12 is very important for the human body. Early diagnosis of vitamin B12 deficiency is essential because it is associated with many problems such as fatigue, lethargy, depression, poor memory, breathlessness, headaches, pale skin, mania, psychosis, etc. Most of
Alexis Loiseau et al.
Cancers, 11(12) (2019-12-11)
Nanohybrids based on titanate nanotubes (TiONts) were developed to fight prostate cancer by intratumoral (IT) injection, and particular attention was paid to their step-by-step synthesis. TiONts were synthesized by a hydrothermal process. To develop the customengineered nanohybrids, the surface of
Colin R Zamecnik et al.
ACS nano, 11(11), 11433-11440 (2017-11-11)
Injectable nanomaterials that interact with the host immune system without surgical intervention present spatially anchored complements to cell transplantation and could offer improved pharmacokinetics compared to systemic cytokine therapy. Here we demonstrate fabrication of high aspect ratio polycaprolactone nanowires coupled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service