Skip to Content
Merck
All Photos(1)

Key Documents

562882

Sigma-Aldrich

4-Propylbenzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H4CHO
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.532 (lit.)

density

1.0050 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(CCC)cc1

InChI

1S/C10H12O/c1-2-3-9-4-6-10(8-11)7-5-9/h4-8H,2-3H2,1H3

InChI key

MAUCRURSQMOFGV-UHFFFAOYSA-N

Application

4-Propylbenzaldehyde may be used in the synthesis of the following:
  • (E)-N-(4-propylbenzylidene)-2-((4-methoxyphenyl)amino)acetohydrazide
  • 2-(4-propylbenzylidene)-1-(5-bromo-2 morpholinopyrimidin-4-yl)hydrazine
  • H2-5(p-carboxyphenyl)-10,15,20-tri-(4-propyl)phenylporphyrin
  • 3-(4-propylphenyl)propanenitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of (E)-2-(arylbenzylidene)-2-((4-methoxyphenyl) amino) acetohydrazide derivatives and their antimicrobial activity"
Kumar PNB, et al.
Current Chemistry Letters, 2(4), 167-176 (2013)
"Functionalization of Diameter-Sorted Semiconductive SWCNTs with Photosensitizing Porphyrins: Syntheses and Photoinduced Electron Transfer"
Das KS, et al.
Chemistry?A European Journal , 18(36), 11388-11398 (2012)
Kraft P and Swift.D.AK et al.
Perspectives in Flavor and Fragrance Research (2005)
Synthesis of new 1, 2, 4-triazole derivatives and their anticorrosion properties on mild steel in hydrochloric acid medium.
Gurudatt DM, et al.
Journal of Molecular Liquids, 211, 275-287 (2015)
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service