Skip to Content
Merck
All Photos(2)

Key Documents

560391

Sigma-Aldrich

PSS-Trisilanol-isooctyl substituted

Synonym(s):

1,3,5,7,9,11,14-Hepta-isooctyltricyclo[7.3.3.15,11]heptasiloxane-endo-3,7,14-triol, Trisilanolisooctyl-POSS®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H122O12Si7
CAS Number:
Molecular Weight:
1184.16
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

refractive index

n20/D 1.4550 (lit.)

solubility

THF: soluble
acetone: soluble
acetonitrile: insoluble
chloroform: soluble
ethanol: soluble
hexane: soluble

density

0.969 g/mL at 25 °C (lit.)

SMILES string

[Si]21(O[Si]3(O[Si](O[Si](O2)(O[Si](O[Si](O1)(O[Si](O3)(O)CCCCCC(C)C)CCCCCC(C)C)(O)CCCCCC(C)C)CCCCCC(C)C)(O)CCCCCC(C)C)CCCCCC(C)C)CCCCCC(C)C

InChI

1S/C56H122O12Si7/c1-50(2)36-22-15-29-43-69(57)60-72(46-32-18-25-39-53(7)8)62-70(58,44-30-16-23-37-51(3)4)64-74(48-34-20-27-41-55(11)12)65-71(59,45-31-17-24-38-52(5)6)63-73(61-69,47-33-19-26-40-54(9)10)67-75(66-72,68-74)49-35-21-28-42-56(13)14/h50-59H,15-4

InChI key

DKOTXLDMWAXTTD-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.9 °F - closed cup

Flash Point(C)

68.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service