560375
PSS-(3-Mercapto)propyl-Heptaisobutyl substituted
Synonym(s):
1-(3-Mercapto)propyl-3,5,7,9,11,13,15-isobutylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Mercaptopropylisobutyl-POSS®, POSS®-Mercaptopropyl-isobutyl substituted
About This Item
Recommended Products
mp
230-235 °C (lit.)
Quality Level
SMILES string
CC(C)C[Si]12O[Si]3(CCCS)O[Si]4(CC(C)C)O[Si](CC(C)C)(O1)O[Si]5(CC(C)C)O[Si](CC(C)C)(O2)O[Si](CC(C)C)(O3)O[Si](CC(C)C)(O4)O5
InChI
1S/C31H70O12SSi8/c1-25(2)18-46-32-45(17-15-16-44)33-47(19-26(3)4)37-49(35-46,21-28(7)8)41-52(24-31(13)14)42-50(36-46,22-29(9)10)38-48(34-45,20-27(5)6)40-51(39-47,43-52)23-30(11)12/h25-31,44H,15-24H2,1-14H3
InChI key
CYGQPMMZQIQYKC-UHFFFAOYSA-N
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service